Sonoco ThermoSafe Insulated Shippers
EPS Insulated Shippers
ThermoSafe™ offers a line of EPS insulated shippers that are the ideal solution for the safe storage and delivery of most temperature-sensitive products. Shippers available as foam-only or in corrugated cartons. Standard wall thickness, 1.5 inches. EPS shippers are ideal for one- and two-day shipments.
PUR Insulated Shippers
ThermoSafe high-performance polyurethane containers meet the needs of the most demanding temperature-sensitive payloads. Polyurethane insulation offers greater insulating power than equivalent thicknesses of EPS containers. Thick walls and 200 lb. outer corrugate offer protective shock absorption and cushioning during shipping and handling. PUR shippers are ideal for two- and three-day shipments.
VIP Insulated Shippers
ThermoSafe high-performance VIP insulated shippers are ideal for longer duration shipments of high-value, temperature-sensitive products. VIP insulated shippers extend shipping duration while requiring less refrigerant to hold temperature. VIP insulated shippers yield in smaller, lighter packages and are ideal for three- to five-day shipments.