Assay Details
Gene Symbol: LOC5572804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846995.1 | XP_021702687.1 | 3 - 4 | 1954 | 76 | |
XM_021846997.1 | XP_021702689.1 | 3 - 4 | 1954 | 76 | ||
XM_021846998.1 | XP_021702690.1 | 3 - 4 | 2215 | 76 | ||
XM_021846999.1 | XP_021702691.1 | 3 - 4 | 1909 | 76 | ||
XM_021847000.1 | XP_021702692.1 | 3 - 4 | 2392 | 76 | ||
XM_021847001.1 | XP_021702693.1 | 3 - 4 | 1825 | 76 | ||
XM_021847002.1 | XP_021702694.1 | 3 - 4 | 1958 | 76 |
