Assay Details
Gene Symbol: LOC5572804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846995.1 | XP_021702687.1 | 4 - 5 | 2864 | 105 | |
XM_021846997.1 | XP_021702689.1 | 4 - 5 | 2864 | 105 | ||
XM_021846998.1 | XP_021702690.1 | 4 - 5 | 3125 | 105 | ||
XM_021846999.1 | XP_021702691.1 | 4 - 5 | 2819 | 105 | ||
XM_021847000.1 | XP_021702692.1 | 4 - 5 | 3302 | 105 | ||
XM_021847001.1 | XP_021702693.1 | 4 - 5 | 2735 | 105 | ||
XM_021847002.1 | XP_021702694.1 | 4 - 5 | 2868 | 105 |

More Information
Set Membership: |
![]() ![]() |
