Assay Details
Gene Symbol: LOC5568181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848214.1 | XP_021703906.1 | 8 - 9 | 2149 | 137 | |
XM_021848215.1 | XP_021703907.1 | 7 - 8 | 2499 | 137 | ||
XM_021848216.1 | XP_021703908.1 | 8 - 9 | 2431 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() |
