Assay Details
Gene Symbol: LOC5570087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840893.1 | XP_021696585.1 | 1 - 2 | 1297 | 78 | |
XM_021840895.1 | XP_021696587.1 | 1 - 2 | 1297 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021840893.1, XM_021840895.1 transcripts.
Set Membership: |
![]() ![]() |
