Assay Details
Gene Symbol: LOC5570087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840893.1 | XP_021696585.1 | 5 - 6 | 2749 | 62 | |
XM_021840895.1 | XP_021696587.1 | 5 - 6 | 2731 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
