Assay Details
Gene Symbol: LOC5573626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849854.1 | XP_021705546.1 | 10 - 11 | 2808 | 67 | |
XM_021849860.1 | XP_021705552.1 | 10 - 11 | 2805 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
