Assay Details
Gene Symbol: LOC5573626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849854.1 | XP_021705546.1 | 9 - 10 | 2505 | 84 | |
XM_021849860.1 | XP_021705552.1 | 9 - 10 | 2502 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
