Assay Details
Gene Symbol: LOC5579912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848823.1 | XP_021704515.1 | 10 - 11 | 3387 | 77 | |
XM_021848824.1 | XP_021704516.1 | 10 - 11 | 3502 | 77 | ||
XM_021848825.1 | XP_021704517.1 | 10 - 11 | 3441 | 77 | ||
XM_021848826.1 | XP_021704518.1 | 11 - 12 | 3752 | 77 | ||
XM_021848827.1 | XP_021704519.1 | 10 - 11 | 3406 | 77 | ||
XM_021848828.1 | XP_021704520.1 | 10 - 11 | 3354 | 77 | ||
XM_021848830.1 | XP_021704522.1 | 10 - 11 | 3326 | 77 | ||
XM_021848831.1 | XP_021704523.1 | 10 - 11 | 3323 | 77 | ||
XM_021848832.1 | XP_021704524.1 | 8 - 9 | 2026 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
