Assay Details
Gene Symbol: LOC5579912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848824.1 | XP_021704516.1 | 2 - 3 | 1310 | 60 | |
XM_021848825.1 | XP_021704517.1 | 2 - 3 | 1249 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848824.1, XM_021848825.1 transcripts.
Set Membership: |
![]() ![]() |
