Assay Details
Gene Symbol: LOC5579912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848823.1 | XP_021704515.1 | 5 - 6 | 1826 | 71 | |
XM_021848824.1 | XP_021704516.1 | 5 - 6 | 1941 | 71 | ||
XM_021848825.1 | XP_021704517.1 | 5 - 6 | 1880 | 71 | ||
XM_021848826.1 | XP_021704518.1 | 6 - 7 | 2191 | 71 | ||
XM_021848827.1 | XP_021704519.1 | 5 - 6 | 1845 | 71 | ||
XM_021848828.1 | XP_021704520.1 | 5 - 6 | 1793 | 71 | ||
XM_021848830.1 | XP_021704522.1 | 5 - 6 | 1765 | 71 | ||
XM_021848831.1 | XP_021704523.1 | 5 - 6 | 1762 | 71 | ||
XM_021848832.1 | XP_021704524.1 | 3 - 4 | 465 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
