Assay Details
Gene Symbol: LOC5579912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848823.1 | XP_021704515.1 | 6 - 7 | 1977 | 73 | |
XM_021848824.1 | XP_021704516.1 | 6 - 7 | 2092 | 73 | ||
XM_021848825.1 | XP_021704517.1 | 6 - 7 | 2031 | 73 | ||
XM_021848826.1 | XP_021704518.1 | 7 - 8 | 2342 | 73 | ||
XM_021848827.1 | XP_021704519.1 | 6 - 7 | 1996 | 73 | ||
XM_021848828.1 | XP_021704520.1 | 6 - 7 | 1944 | 73 | ||
XM_021848830.1 | XP_021704522.1 | 6 - 7 | 1916 | 73 | ||
XM_021848831.1 | XP_021704523.1 | 6 - 7 | 1913 | 73 | ||
XM_021848832.1 | XP_021704524.1 | 4 - 5 | 616 | 73 |
