Assay Details
Gene Symbol: LOC5579912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848823.1 | XP_021704515.1 | 8 - 9 | 2772 | 79 | |
XM_021848824.1 | XP_021704516.1 | 8 - 9 | 2887 | 79 | ||
XM_021848825.1 | XP_021704517.1 | 8 - 9 | 2826 | 79 | ||
XM_021848826.1 | XP_021704518.1 | 9 - 10 | 3137 | 79 | ||
XM_021848827.1 | XP_021704519.1 | 8 - 9 | 2791 | 79 | ||
XM_021848828.1 | XP_021704520.1 | 8 - 9 | 2739 | 79 | ||
XM_021848830.1 | XP_021704522.1 | 8 - 9 | 2711 | 79 | ||
XM_021848831.1 | XP_021704523.1 | 8 - 9 | 2708 | 79 | ||
XM_021848832.1 | XP_021704524.1 | 6 - 7 | 1411 | 79 |
