Assay Details
Gene Symbol: LOC5579912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848823.1 | XP_021704515.1 | 9 - 10 | 3229 | 73 | |
XM_021848824.1 | XP_021704516.1 | 9 - 10 | 3344 | 73 | ||
XM_021848825.1 | XP_021704517.1 | 9 - 10 | 3283 | 73 | ||
XM_021848826.1 | XP_021704518.1 | 10 - 11 | 3594 | 73 | ||
XM_021848827.1 | XP_021704519.1 | 9 - 10 | 3248 | 73 | ||
XM_021848828.1 | XP_021704520.1 | 9 - 10 | 3196 | 73 | ||
XM_021848830.1 | XP_021704522.1 | 9 - 10 | 3168 | 73 | ||
XM_021848831.1 | XP_021704523.1 | 9 - 10 | 3165 | 73 | ||
XM_021848832.1 | XP_021704524.1 | 7 - 8 | 1868 | 73 |
