Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842723.1 | XP_021698415.1 | 2 - 3 | 909 | 85 | |
XM_021842724.1 | XP_021698416.1 | 3 - 4 | 1519 | 85 | ||
XM_021842725.1 | XP_021698417.1 | 3 - 4 | 1515 | 85 | ||
XM_021842726.1 | XP_021698418.1 | 3 - 4 | 1493 | 85 | ||
XM_021842727.1 | XP_021698419.1 | 3 - 4 | 1489 | 85 | ||
XM_021842728.1 | XP_021698420.1 | 2 - 3 | 1369 | 85 | ||
XM_021842729.1 | XP_021698421.1 | 2 - 3 | 1368 | 85 | ||
XM_021842733.1 | XP_021698425.1 | 2 - 3 | 1369 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842723.1, XM_021842724.1, XM_021842725.1, XM_021842726.1, XM_021842727.1, XM_021842728.1, XM_021842729.1, XM_021842733.1 transcripts.
Set Membership: |
![]() ![]() |
