Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842728.1 | XP_021698420.1 | 1 - 2 | 552 | 131 | |
XM_021842729.1 | XP_021698421.1 | 1 - 2 | 551 | 131 | ||
XM_021842733.1 | XP_021698425.1 | 1 - 2 | 552 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842728.1, XM_021842729.1, XM_021842733.1 transcripts.
Set Membership: |
![]() ![]() |
