Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842723.1 | XP_021698415.1 | 15 - 16 | 9441 | 76 | |
XM_021842724.1 | XP_021698416.1 | 16 - 17 | 10051 | 76 | ||
XM_021842725.1 | XP_021698417.1 | 16 - 17 | 10047 | 76 | ||
XM_021842726.1 | XP_021698418.1 | 16 - 17 | 10025 | 76 | ||
XM_021842727.1 | XP_021698419.1 | 16 - 17 | 10021 | 76 | ||
XM_021842728.1 | XP_021698420.1 | 14 - 15 | 9898 | 76 | ||
XM_021842729.1 | XP_021698421.1 | 14 - 15 | 9276 | 76 | ||
XM_021842730.1 | XP_021698422.1 | 13 - 14 | 5017 | 76 | ||
XM_021842731.1 | XP_021698423.1 | 12 - 13 | 4806 | 76 | ||
XM_021842732.1 | XP_021698424.1 | 12 - 13 | 4611 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
