Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842723.1 | XP_021698415.1 | 13 - 14 | 8902 | 79 | |
XM_021842724.1 | XP_021698416.1 | 14 - 15 | 9512 | 79 | ||
XM_021842725.1 | XP_021698417.1 | 14 - 15 | 9508 | 79 | ||
XM_021842726.1 | XP_021698418.1 | 14 - 15 | 9486 | 79 | ||
XM_021842727.1 | XP_021698419.1 | 14 - 15 | 9482 | 79 | ||
XM_021842728.1 | XP_021698420.1 | 12 - 13 | 9359 | 79 | ||
XM_021842729.1 | XP_021698421.1 | 12 - 13 | 8737 | 79 | ||
XM_021842730.1 | XP_021698422.1 | 11 - 12 | 4478 | 79 | ||
XM_021842731.1 | XP_021698423.1 | 10 - 11 | 4267 | 79 | ||
XM_021842732.1 | XP_021698424.1 | 10 - 11 | 4072 | 79 | ||
XM_021842733.1 | XP_021698425.1 | 12 - 13 | 9359 | 79 |
