Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842728.1 | XP_021698420.1 | 11 - 12 | 8541 | 103 | |
XM_021842731.1 | XP_021698423.1 | 9 - 10 | 3449 | 103 | ||
XM_021842733.1 | XP_021698425.1 | 11 - 12 | 8541 | 103 |

More Information
Set Membership: |
![]() ![]() |
