Assay Details
Gene Symbol: LOC5576159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662454.2 | XP_001662504.2 | 5 - 6 | 1220 | 80 | |
XM_021853637.1 | XP_021709329.1 | 4 - 5 | 809 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
