Assay Details
Gene Symbol: LOC5568333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839256.1 | XP_021694948.1 | 2 - 3 | 858 | 85 | |
XM_021839257.1 | XP_021694949.1 | 1 - 2 | 1187 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839256.1, XM_021839257.1 transcripts.
Set Membership: |
![]() ![]() |
