Assay Details
Gene Symbol: LOC5568333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839256.1 | XP_021694948.1 | 3 - 4 | 1392 | 71 | |
XM_021839257.1 | XP_021694949.1 | 2 - 3 | 1721 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
