Assay Details
Gene Symbol: LOC5568333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839256.1 | XP_021694948.1 | 4 - 5 | 1662 | 77 | |
XM_021839257.1 | XP_021694949.1 | 3 - 4 | 1991 | 77 |
