Assay Details
Gene Symbol: LOC5577014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846939.1 | XP_021702631.1 | 8 - 9 | 2371 | 58 | |
XM_021846940.1 | XP_021702632.1 | 8 - 9 | 2370 | 58 |

More Information
Set Membership: |
![]() ![]() |
