Assay Details
Gene Symbol: LOC5577014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846935.1 | XP_021702627.1 | 9 - 10 | 3131 | 75 | |
XM_021846936.1 | XP_021702628.1 | 9 - 10 | 3063 | 75 | ||
XM_021846937.1 | XP_021702629.1 | 9 - 10 | 3116 | 75 | ||
XM_021846939.1 | XP_021702631.1 | 10 - 11 | 2990 | 75 | ||
XM_021846940.1 | XP_021702632.1 | 10 - 11 | 2989 | 75 |
