Assay Details
Gene Symbol: LOC5570771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659311.2 | XP_001659361.1 | 3 - 4 | 2422 | 63 | |
XM_021838239.1 | XP_021693931.1 | 3 - 4 | 2425 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
