Assay Details
Gene Symbol: LOC5570771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659311.2 | XP_001659361.1 | 2 - 3 | 2219 | 77 | |
XM_021838239.1 | XP_021693931.1 | 2 - 3 | 2222 | 77 |
