Assay Details
Gene Symbol: LOC5566523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845427.1 | XP_021701119.1 | 9 - 10 | 1947 | 82 | |
XM_021845429.1 | XP_021701121.1 | 9 - 10 | 1957 | 82 | ||
XM_021845430.1 | XP_021701122.1 | 9 - 10 | 1866 | 82 | ||
XM_021845431.1 | XP_021701123.1 | 9 - 10 | 1933 | 82 | ||
XM_021845432.1 | XP_021701124.1 | 9 - 10 | 1762 | 82 | ||
XM_021845433.1 | XP_021701125.1 | 9 - 10 | 1876 | 82 | ||
XM_021845434.1 | XP_021701126.1 | 9 - 10 | 1852 | 82 | ||
XM_021845435.1 | XP_021701127.1 | 9 - 10 | 1815 | 82 | ||
XM_021845436.1 | XP_021701128.1 | 9 - 10 | 1772 | 82 | ||
XM_021845437.1 | XP_021701129.1 | 9 - 10 | 1748 | 82 | ||
XM_021845438.1 | XP_021701130.1 | 9 - 10 | 1882 | 82 | ||
XM_021845440.1 | XP_021701132.1 | 9 - 10 | 1801 | 82 | ||
XM_021845441.1 | XP_021701133.1 | 9 - 10 | 1697 | 82 | ||
XM_021845442.1 | XP_021701134.1 | 9 - 10 | 1764 | 82 | ||
XM_021845443.1 | XP_021701135.1 | 8 - 9 | 1749 | 82 | ||
XM_021845444.1 | XP_021701136.1 | 9 - 10 | 1579 | 82 | ||
XM_021845445.1 | XP_021701137.1 | 7 - 8 | 1360 | 82 | ||
XM_021845446.1 | XP_021701138.1 | 9 - 10 | 1947 | 82 | ||
XM_021845447.1 | XP_021701139.1 | 9 - 10 | 1947 | 82 |
