Assay Details
Gene Symbol: LOC5566523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845427.1 | XP_021701119.1 | 6 - 7 | 1445 | 59 | |
XM_021845429.1 | XP_021701121.1 | 6 - 7 | 1455 | 59 | ||
XM_021845430.1 | XP_021701122.1 | 6 - 7 | 1364 | 59 | ||
XM_021845431.1 | XP_021701123.1 | 6 - 7 | 1431 | 59 | ||
XM_021845432.1 | XP_021701124.1 | 6 - 7 | 1260 | 59 | ||
XM_021845433.1 | XP_021701125.1 | 6 - 7 | 1374 | 59 | ||
XM_021845434.1 | XP_021701126.1 | 6 - 7 | 1350 | 59 | ||
XM_021845435.1 | XP_021701127.1 | 6 - 7 | 1313 | 59 | ||
XM_021845436.1 | XP_021701128.1 | 6 - 7 | 1270 | 59 | ||
XM_021845437.1 | XP_021701129.1 | 6 - 7 | 1246 | 59 | ||
XM_021845438.1 | XP_021701130.1 | 6 - 7 | 1380 | 59 | ||
XM_021845440.1 | XP_021701132.1 | 6 - 7 | 1299 | 59 | ||
XM_021845441.1 | XP_021701133.1 | 6 - 7 | 1195 | 59 | ||
XM_021845442.1 | XP_021701134.1 | 6 - 7 | 1262 | 59 | ||
XM_021845443.1 | XP_021701135.1 | 5 - 6 | 1247 | 59 | ||
XM_021845444.1 | XP_021701136.1 | 6 - 7 | 1077 | 59 | ||
XM_021845445.1 | XP_021701137.1 | 4 - 5 | 858 | 59 | ||
XM_021845446.1 | XP_021701138.1 | 6 - 7 | 1445 | 59 | ||
XM_021845447.1 | XP_021701139.1 | 6 - 7 | 1445 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
