Assay Details
Gene Symbol: LOC5566523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845427.1 | XP_021701119.1 | 4 - 5 | 1225 | 67 | |
XM_021845429.1 | XP_021701121.1 | 4 - 5 | 1235 | 67 | ||
XM_021845430.1 | XP_021701122.1 | 4 - 5 | 1144 | 67 | ||
XM_021845431.1 | XP_021701123.1 | 4 - 5 | 1211 | 67 | ||
XM_021845432.1 | XP_021701124.1 | 4 - 5 | 1040 | 67 | ||
XM_021845433.1 | XP_021701125.1 | 4 - 5 | 1154 | 67 | ||
XM_021845434.1 | XP_021701126.1 | 4 - 5 | 1130 | 67 | ||
XM_021845435.1 | XP_021701127.1 | 4 - 5 | 1093 | 67 | ||
XM_021845436.1 | XP_021701128.1 | 4 - 5 | 1050 | 67 | ||
XM_021845437.1 | XP_021701129.1 | 4 - 5 | 1026 | 67 | ||
XM_021845438.1 | XP_021701130.1 | 4 - 5 | 1160 | 67 | ||
XM_021845440.1 | XP_021701132.1 | 4 - 5 | 1079 | 67 | ||
XM_021845441.1 | XP_021701133.1 | 4 - 5 | 975 | 67 | ||
XM_021845442.1 | XP_021701134.1 | 4 - 5 | 1042 | 67 | ||
XM_021845443.1 | XP_021701135.1 | 3 - 4 | 1027 | 67 | ||
XM_021845444.1 | XP_021701136.1 | 4 - 5 | 857 | 67 | ||
XM_021845445.1 | XP_021701137.1 | 2 - 3 | 638 | 67 | ||
XM_021845446.1 | XP_021701138.1 | 4 - 5 | 1225 | 67 | ||
XM_021845447.1 | XP_021701139.1 | 4 - 5 | 1225 | 67 |

More Information
Set Membership: |
![]() ![]() |
