Assay Details
Gene Symbol: LOC5568336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839258.1 | XP_021694950.1 | 5 - 6 | 714 | 78 | |
XM_021839259.1 | XP_021694951.1 | 6 - 7 | 1451 | 78 | ||
XM_021839260.1 | XP_021694952.1 | 4 - 5 | 438 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
