Assay Details
Gene Symbol: LOC5568336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839258.1 | XP_021694950.1 | 4 - 5 | 615 | 92 | |
XM_021839259.1 | XP_021694951.1 | 5 - 6 | 1352 | 92 | ||
XM_021839260.1 | XP_021694952.1 | 3 - 4 | 339 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
