Assay Details
Gene Symbol: LOC5568336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839258.1 | XP_021694950.1 | 2 - 3 | 311 | 80 | |
XM_021839259.1 | XP_021694951.1 | 3 - 4 | 1048 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
