Assay Details
Gene Symbol: LOC5566011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843177.1 | XP_021698869.1 | 1 - 2 | 478 | 72 | |
XM_021843178.1 | XP_021698870.1 | 1 - 2 | 478 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843177.1, XM_021843178.1 transcripts.
Set Membership: |
![]() ![]() |
