Assay Details
Gene Symbol: LOC5566011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843177.1 | XP_021698869.1 | 2 - 3 | 822 | 66 | |
XM_021843178.1 | XP_021698870.1 | 2 - 3 | 822 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
