Assay Details
Gene Symbol: LOC5563690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853885.1 | XP_021709577.1 | 11 - 12 | 3272 | 104 | |
XM_021853886.1 | XP_021709578.1 | 9 - 10 | 3184 | 104 | ||
XM_021853887.1 | XP_021709579.1 | 11 - 12 | 3230 | 104 | ||
XM_021853888.1 | XP_021709580.1 | 11 - 12 | 3165 | 104 | ||
XM_021853889.1 | XP_021709581.1 | 11 - 12 | 3139 | 104 | ||
XM_021853891.1 | XP_021709583.1 | 11 - 12 | 3109 | 104 | ||
XM_021853892.1 | XP_021709584.1 | 10 - 11 | 2503 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
