Assay Details
Gene Symbol: LOC5563690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853885.1 | XP_021709577.1 | 5 - 6 | 1697 | 93 | |
XM_021853886.1 | XP_021709578.1 | 3 - 4 | 1609 | 93 | ||
XM_021853887.1 | XP_021709579.1 | 5 - 6 | 1655 | 93 | ||
XM_021853888.1 | XP_021709580.1 | 5 - 6 | 1590 | 93 | ||
XM_021853889.1 | XP_021709581.1 | 5 - 6 | 1564 | 93 | ||
XM_021853891.1 | XP_021709583.1 | 5 - 6 | 1534 | 93 | ||
XM_021853892.1 | XP_021709584.1 | 4 - 5 | 928 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
