Assay Details
Gene Symbol: LOC5563690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853885.1 | XP_021709577.1 | 6 - 7 | 1816 | 86 | |
XM_021853886.1 | XP_021709578.1 | 4 - 5 | 1728 | 86 | ||
XM_021853887.1 | XP_021709579.1 | 6 - 7 | 1774 | 86 | ||
XM_021853888.1 | XP_021709580.1 | 6 - 7 | 1709 | 86 | ||
XM_021853889.1 | XP_021709581.1 | 6 - 7 | 1683 | 86 | ||
XM_021853891.1 | XP_021709583.1 | 6 - 7 | 1653 | 86 | ||
XM_021853892.1 | XP_021709584.1 | 5 - 6 | 1047 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
