Assay Details
Gene Symbol: LOC5563690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853885.1 | XP_021709577.1 | 9 - 10 | 3001 | 77 | |
XM_021853886.1 | XP_021709578.1 | 7 - 8 | 2913 | 77 | ||
XM_021853887.1 | XP_021709579.1 | 9 - 10 | 2959 | 77 | ||
XM_021853888.1 | XP_021709580.1 | 9 - 10 | 2894 | 77 | ||
XM_021853889.1 | XP_021709581.1 | 9 - 10 | 2868 | 77 | ||
XM_021853891.1 | XP_021709583.1 | 9 - 10 | 2838 | 77 | ||
XM_021853892.1 | XP_021709584.1 | 8 - 9 | 2232 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
