Assay Details
Gene Symbol: LOC5563721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837523.1 | XP_021693215.1 | 3 - 4 | 920 | 63 | |
XM_021837524.1 | XP_021693216.1 | 3 - 4 | 922 | 63 | ||
XM_021837525.1 | XP_021693217.1 | 3 - 4 | 1004 | 63 |

More Information
Set Membership: |
![]() ![]() |
