Assay Details
Gene Symbol: LOC5569559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839490.1 | XP_021695182.1 | 2 - 3 | 1206 | 82 | |
XM_021839502.1 | XP_021695194.1 | 2 - 3 | 1206 | 82 | ||
XM_021839508.1 | XP_021695200.1 | 2 - 3 | 1206 | 82 | ||
XM_021839511.1 | XP_021695203.1 | 2 - 3 | 1206 | 82 | ||
XM_021839515.1 | XP_021695207.1 | 2 - 3 | 1206 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839490.1, XM_021839502.1, XM_021839508.1, XM_021839511.1, XM_021839515.1 transcripts.
Set Membership: |
![]() ![]() |
