Assay Details
Gene Symbol: LOC5569559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839490.1 | XP_021695182.1 | 6 - 7 | 2598 | 84 | |
XM_021839496.1 | XP_021695188.1 | 5 - 6 | 1696 | 84 | ||
XM_021839502.1 | XP_021695194.1 | 6 - 7 | 2598 | 84 | ||
XM_021839508.1 | XP_021695200.1 | 6 - 7 | 2598 | 84 | ||
XM_021839511.1 | XP_021695203.1 | 6 - 7 | 2598 | 84 | ||
XM_021839515.1 | XP_021695207.1 | 6 - 7 | 2598 | 84 |
