Assay Details
Gene Symbol: LOC5579800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848972.1 | XP_021704664.1 | 11 - 12 | 4364 | 74 | |
XM_021848986.1 | XP_021704678.1 | 11 - 12 | 4351 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
