Assay Details
Gene Symbol: LOC5579800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848978.1 | XP_021704670.1 | 2 - 3 | 784 | 87 | |
XM_021848979.1 | XP_021704671.1 | 1 - 2 | 982 | 87 | ||
XM_021848980.1 | XP_021704672.1 | 2 - 3 | 737 | 87 | ||
XM_021848981.1 | XP_021704673.1 | 2 - 3 | 714 | 87 | ||
XM_021848982.1 | XP_021704674.1 | 2 - 3 | 782 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848978.1, XM_021848979.1, XM_021848980.1, XM_021848981.1, XM_021848982.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
