Assay Details
Gene Symbol: LOC5579800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848960.1 | XP_021704652.1 | 8 - 9 | 3834 | 68 | |
XM_021848961.1 | XP_021704653.1 | 8 - 9 | 3820 | 68 | ||
XM_021848966.1 | XP_021704658.1 | 8 - 9 | 3809 | 68 | ||
XM_021848976.1 | XP_021704668.1 | 8 - 9 | 2843 | 68 | ||
XM_021848982.1 | XP_021704674.1 | 9 - 10 | 2873 | 68 | ||
XM_021848984.1 | XP_021704676.1 | 8 - 9 | 3834 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
