Assay Details
Gene Symbol: LOC5578793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657113.2 | XP_001657163.2 | 1 - 2 | 267 | 64 | |
XM_001657114.2 | XP_001657164.2 | 1 - 2 | 267 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
