Assay Details
Gene Symbol: LOC5576834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852650.1 | XP_021708342.1 | 4 - 5 | 1995 | 107 | |
XM_021852651.1 | XP_021708343.1 | 4 - 5 | 1591 | 107 | ||
XM_021852652.1 | XP_021708344.1 | 4 - 5 | 1844 | 107 | ||
XM_021852653.1 | XP_021708345.1 | 4 - 5 | 1576 | 107 | ||
XM_021852654.1 | XP_021708346.1 | 4 - 5 | 1459 | 107 |

More Information
Set Membership: |
![]() ![]() |
