Assay Details
Gene Symbol: LOC5576748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656199.2 | XP_001656249.1 | 2 - 3 | 579 | 91 | |
XM_021848932.1 | XP_021704624.1 | 2 - 3 | 849 | 91 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5576748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656199.2 | XP_001656249.1 | 2 - 3 | 579 | 91 | |
XM_021848932.1 | XP_021704624.1 | 2 - 3 | 849 | 91 |