Assay Details
Gene Symbol: LOC5576748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656199.2 | XP_001656249.1 | 4 - 5 | 2166 | 106 | |
XM_021848932.1 | XP_021704624.1 | 4 - 5 | 2436 | 106 |

More Information
Set Membership: |
![]() ![]() |
