Assay Details
Gene Symbol: LOC5576748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656199.2 | XP_001656249.1 | 5 - 6 | 2331 | 62 | |
XM_021848932.1 | XP_021704624.1 | 5 - 6 | 2601 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
